missing translation for 'onlineSavingsMsg'
Learn More

XRCC4 Antibody (OTI4H9), Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP1-48053

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215261

  • 572.34€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

XRCC4 Monoclonal specifically detects XRCC4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

XRCC4
Monoclonal
1 mg/mL
Western Blot 1:2000, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin 1:50
XRCC4
Full-length protein expressed in 293T cell transfected with human XRCC4 expression vector
Affinity Purified
RUO
Primary
This antibody is specific for Homo sapiens X-ray repair complementing defective repair in Chinese hamster cells 4 (XRCC4), transcript variant 3.
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
OTI4H9
Unconjugated
complementing defective, repair in Chinese hamster, X-ray repair complementing defective repair in Chinese hamster cells 4, X-ray repair cross-complementing protein 4
Mouse
38.3 kDa
0.1 mL
Cancer, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Tumor Suppressors
7518
Human
IgG1
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only