Learn More
Thermo Scientific™ Xpect™ Cryptosporidium Test
Product Code. 13215629
Change view
Click to view available options
Quantity:
20 Tests/Kit
Unit Size:
20 tests
Product Code. Quantity Unit Size
13215629 20 Tests/Kit 20 tests
1 options
This item is not returnable. View return policy
Product Code. 13215629 Supplier Thermo Scientific™ Supplier No. R2451020
  • ONLINE EXCLUSIVE PRICE
    483.00€
    293.93€ / 20 tests
    Ahorro 189.07€
    39% Descuento
 Restricted Item
Fecha estimada de envoi: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Rapidly detect Cryptosporidium-specific antigens from fresh, frozen, or preserved human fecal samples.
Optimize your workflow by rapidly detecting Cryptosporidium-specific antigen from fresh, frozen, or preserved human fecal specimens using Thermo Scientific™ Xpect™ Cryptosporidium Test. Report accurate results in 15 minutes during all stages of infection—all from a single specimen.
Use this rapid, lateral flow immunoassay for the direct qualitative detection of Cryptosporidium-specific antigen. Enhance laboratory performance and efficiency with excellent clinical sensitivity and specificity and a simple walk-away procedure.
  • Clearly visible, easy-to-read results in just 15 minutes
  • Less than one minute hands-on time
  • 95.8 to 97.9% sensitivity
  • Single and combination immunoassays available
  • No specimen dilution required when using the recommended Formalin, SAF, or modified Cary-Blair w/Indicator transport media
Xpect Cryptosporidium uses antibody-coated color particles to bind with target antigen in the diluted sample. The color-antibody/antigen complex moves along the test membrane. Immobilised antibodies specific to target antigen capture the color-labelled antigen complex at the test region, causing a clearly visible line to appear. This line will not appear in the absence of specific antigen. Immobilised, non-specific antibodies in the control region capture the color-labelled antibodies, whether antigen is present or not, resulting in a clearly visible control line.
Trust the entire family of Xpect rapid tests to deliver unparalleled diagnostic performance while offering ease-of-use, convenience and time savings. Each test is an immunochromatographic assay with a simple procedure that provides rapid, cost-effective detection to aid in the diagnosis of infections.
Xpect Flu A&B
Xpect RSV
Xpect C. difficile Toxin A/B
Xpect Rotavirus
Xpect Giardia
Xpect Cryptosporidium
Xpect Giardia/Cryptosporidium
Xpect Legionella
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C
Description Xpect Cryptosporidium Test
Includes Test devices, conjugate (2.5mL), specimen dilution buffer (2.5mL), procedure card, disposable transfer pipettes, dilution tubes
Quantity 20 Tests/Kit