missing translation for 'onlineSavingsMsg'
Learn More

XPA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 16462-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16816094

  • 319.00€ / 150 microlitros

Envío estimado: 29-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The XPA (xeroderma pigmentosumgroup A) protein specifically recognizes the UV-orchemically damaged DNA lesions, and triggers thenucleotide excision repair process. XPA binds to thereplication protein A (RPA) or the excision repaircross complementing 1 protein (ERCC 1). In the absence of nucleotide excision repair persisting (unrepaired) DNA lesions (adducts) may lead to the accumulation of gene mutations and ultimately to cancer. Xeroderma pigmentosum patients have a >2000 fold increased risk to develop skin cancer atsun-exposed areas.
Especificaciones

Especificaciones

XPA
0.22 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P23025, Q64267
XP1, XPA, XPAC
Rabbit
IgG
150 μL
-20°C
Primary
22590, 298074, 7507
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
XPA
Liquid
XPA
XPA Fusion Protein Ag9536
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documentos
Certificados