Learn More
Microspec™ Wrappastack™ Starter Kit
Product Code. 16615482
Change view
Click to view available options
Diameter (Metric):
55 mm
Unit Size:
1 set
Product Code. Diameter (Metric) Unit Size
16615482 55 mm 1 set
1 options
This item is not returnable. View return policy
Product Code. 16615482 Supplier Microspec™ Supplier No. K6
  • ONLINE EXCLUSIVE PRICE
    138.00€
    131.52€ / 1 sets
    Ahorro 6.48€
    4% Descuento
Fecha estimada de envoi: 29-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
With 55 mm sleeves
Starter Kit
Specifications
Autoclavable Not Autoclavable
Capacity (Metric) 10 Dishes
Color Clear
Disposable Single-use
Dnase Free No
Diameter (Metric) 55 mm