Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Disposable NMR Tube Caps, 4.0mm Dia., Blue
Different colors help track samples based on various reference solvents.
Marca:  Bel-Art™ 66004-0101
Detalles adicionales : Peso : 0.04310kg
 Ver más versiones de este producto
Código de producto. 15328708
  • 10.10€ / paquete de 100
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
Alert: When using Chloroform or Acetone as a reference solution, please avoid direct contact between the sample and the disposable cap, and avoid long term storage over 2 hours. If these conditions cannot be avoided, please use PTFE tube caps.
Especificación
Especificación
4.0mm
Polyethylene
NMR Spectroscopy
Disposable
Model: Wilmad™ 521-G-100
Blue
100 per pack
Videos