missing translation for 'onlineSavingsMsg'
Learn More

Wilm's Tumor 1 Rabbit, Clone: WT1/1434R, enQuireBio™

Rabbit Monoclonal Antibody

Marca:  EnquireBio 7490-RBM4-P2

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15931098

  • 591.00€ / 200 microgramos


Fecha estimada de envío
21-10-2019
Añadir a la cesta

Especificaciones

Especificaciones

Wilm's Tumor 1
Nuclear and Cytoplasmic
200μg/mL
10mM PBS with or without 0.05% BSA and 0.05% azide.
WT1
Recombinant human full-length WT1 protein
47-55kDa
Protein A/G Purified
RUO
Primary
7490
Immunohistochemistry
WT1/1434R
Unconjugated
NP_001185480
Rabbit
IgG kappa
K562 cells. Wilm s Tumor, mesothelioma or fetal kidney.
200 μg
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Monoclonal
Videos
Documentos