Learn More
Spectrum™ Labs White Polypropylene Universal Closure
Product Code. 11521120
Change view
Click to view available options
Quantity:
10 Pack
Unit Size:
Pack of 10
Product Code. Quantity Unit Size
11521120 10 Pack Pack of 10
1 options
This item is not returnable. View return policy
Product Code. 11521120 Supplier Spectrum™ Labs Supplier No. 142170
  • ONLINE EXCLUSIVE PRICE
    146.00€
    130.88€ / paquete de 10
    Ahorro 15.12€
    10% Descuento
Fecha estimada de envoi: 08-03-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Product Type Universal Closure
Material Polypropylene
Closure Material Polypropylene
Closure Type Universal Closure
Color White
Sterilization Method Gamma Irradiation or Ethylene Oxide
Temperature Range (Metric) 0°C to 90°C
Quantity 10 Pack
For Use With (Application) For standard grade RC dialysis tubing