Learn More
DWK Life Sciences WHEATON™ Snap Plug Caps
Product Code. 12418892 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Blue
Green
Natural
Closure Size:
8 mm
12 mm
15 mm
Unit Size:
Pack of 1000
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Closure Size Unit Size
12418892 Blue 8 mm Pack of 1000
11591245 Natural 12 mm Pack of 1000
12428892 Green 8 mm Pack of 1000
12458892 Natural 15 mm Pack of 1000
12498882 Natural 8 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12418892 Supplier DWK Life Sciences Supplier No. 242804
  • ONLINE EXCLUSIVE PRICE
    104.00€
    94.68€ / paquete de 1000
    Ahorro 9.32€
    8% Descuento
View all promotions
Fecha estimada de envoi: 22-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
WHEATON™ Snap Plug Caps are polyethylene closures designed to provide a secure snap fit for vials and tubes. Available in multiple sizes and colors, these caps feature a TFE liner for reliable sealing in laboratory and clinical applications.
  • Closure style: Snap plug cap for secure, reliable sealing
  • Material: Polyethylene construction for chemical compatibility
  • Liner material: TFE (polytetrafluoroethylene) for enhanced chemical resistance
  • Available in multiple sizes and colors to suit a range of vial and tube formats
  • Suitable for laboratory and clinical use
Specifications
Color Blue
Diameter (Metric) Outer 8 mm
Material LDPE
Sterility Non-sterile
Closure Type Snap Cap
Quantity 1000/Cs.
Closure Size 8 mm
Product Type Plug Cap