Learn More
DWK Life Sciences Wheaton™ Lab 45 Graduated Bottles With Cap Attached
Graduated bottles with cap attached
Marca: DWK Life Sciences 219929
Detalles adicionales : Peso : 4.71740kg
Especificaciones
Polypropylene | |
Screw Cap | |
186 mm | |
17 oz., 500 mL | |
Clear | |
Borosilicate Glass |
45 mm | |
86 mm | |
Wide | |
500 mL | |
Pouring ring and screw cap | |
White |