Learn More
DWK LIFE SCIENCES™ WHEATON™ Deck Kit for Compact Roller System for Mini Bottles
Product Code. 12458952 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Black
Charcoal
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
12458952 Black Each
10563133 Charcoal Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12458952 Supplier DWK LIFE SCIENCES™ Supplier No. 348920
  • ONLINE EXCLUSIVE PRICE
    1075.00€
    817.14€ / paquete de 1
    Ahorro 257.86€
    23% Descuento
Fecha estimada de envoi: 04-03-2027
Add to basket
Add to basket
This item is not returnable. View return policy
The WHEATON™ Deck Kit for Compact Roller System for Mini Bottles expands your Mini Bottle Bench-Top Roller Rack to accommodate an additional 4-12 bottle positions per deck. Up to 2 deck kits can be added per unit.
  • Deck kit designed for use with the Wheaton Mini Bottle Bench-Top Roller Rack
  • Adds 4-12 bottle positions per deck
  • Up to 2 deck kits can be added per unit
Specifications
Product Type Deck Kit
Weight (English) 9 lb.
Weight (Metric) 4 kg
Color Black
For Use With (Equipment) Mini Bottle Roller Culture Apparatus
Includes Assembled roller deck, 4 support posts, drive belt, necessary hardware and assembly instructions