missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Welding Cut Gloves

Heatproof and water-repellent cowhide are combined with cut and thermally resistant Kevlar™ for durability, resistance, and comfort. Honeywell Safety Products™ Welding Cut Gloves are designed with a protective cuff and an elasticized wrist for a good, protective fit.

Marca:  Honeywell Safety Products™ 2049294-09

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 12631113

  • 36.40€ / cada uno


Consúltenos : 902 239 303 
Añadir a la cesta

Descripción

Descripción

  • Suitable for MIG and ARC welding in dry or light oily conditions with cut hazards.
  • Water repellent, heatproof cowhide grain leather back.
  • Water-repellent, heatproof velvet cowhide palm.
  • Water repellent, heatproof split leather cuff to protect the forearm.
  • U.S. assembly.
  • Gunn cut, winged thumb with cord, Crispin type.
  • Elasticated at the back of the hand.
  • Kevlar™ cotton lining for comfort and thermal insulation. Outstanding comfort and flexibility.
Especificaciones

Especificaciones

Cowhide grain leather
Dexterity: 5
Gunn
15cm
Cowhide Leather
2
9
Winged with yoke
Palm and back: 0.8/1.1mm, Cuff: 1.1/1.3mm
Gauntlet, Welding Type A, Crispin
CE CATEGORY II Intermediary risks, NF EN 420-03 EEC Basic standard, NF EN 388-03 Mechanical risks, NF EN 407-04 Thermical risks, EN 12477-02 Type A Welding risks, EC Category PPE 2, EC Certificate Number 0075/797/162/10/12/0763, EC Attestation Number 0075/797/162/10/12/0763
Split leather
2
Automotive, Building, Construction, Iron, Steel, Metal, Ship Building and Welding industries
Dupont™ Kevlar™ aramid fiber, Cotton
Yes
Heat
2
10Pair/PK
Glove
39cm
Videos
Documentos