Descripción
- HDPE coating provides excellent protection against high concentration chemicals, as well as biological risks
- Sealed and coated heat-sealed seams provide splash-proof protection under pressure or in aerosol form
- Elastic tightening under the knee and calf support strap
- Boot height of 50 cm to cover up to the knee to protect the most exposed areas
- Antistatic treated material facilitating the dissipation of electrostatic charges
- Ideal protection against: petroleum products, acids such as nitric or sulfuric, fine powders and wastewater
Especificaciones
Especificaciones
| Color | Yellow |
| Material | Polyethylene |
| Tamaño | Universal |
| Waterproof | Yes |
| Tipo de producto | Overboot |