missing translation for 'onlineSavingsMsg'
Learn More

WBS15/LAT2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11008-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16817783

  • 319.00€ / 150 microlitros

Envío estimado: 02-11-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

NTAL (Non-T cell Activation Linker), also known as LAB (Linker for activation of B cells), is a 30 kDa double-palmitoylated transmembrane adaptor protein expressed by B cells, NK cells, mast cells and macrophages. It is a negative regulator of early stages of BCR-dependent B cell signaling and serves as a negative regulator also in mast cells. However, in mast cells, NTAL also contributes to some activation processes, partially overlapping with LAT function.
Especificaciones

Especificaciones

WBS15/LAT2
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9GZY6, Q9JHL0
HSPC046, LAB, LAT2, Non T cell activation linker, NTAL, WBS15, WBSCR15, WBSCR5, WSCR5
Rabbit
IgG
150 μL
-20°C
Primary
56743, 7462
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
LAT2
Liquid
Lat2
WBS15/LAT2 Fusion Protein Ag1454
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
Documentos
Certificados