Learn More
Rogo Sampaic™ Watch Glasses
Product Code. 11778084
Change view
Click to view available options
Diameter (Metric):
40 mm
50 mm
60 mm
70 mm
80 mm
100 mm
120 mm
140 mm
160 mm
Unit Size:
Pack of 10
Pack of 20
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
11778084 50 mm Pack of 20
11702724 60 mm Pack of 20
11792714 40 mm Pack of 20
11738094 120 mm Pack of 10
11728094 100 mm Pack of 20
11748094 140 mm Pack of 10
11724146 160 mm Pack of 10
11708094 70 mm Pack of 20
11718094 80 mm Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 11778084 Supplier Rogo Sampaic™ Supplier No. KBN012
  • ONLINE EXCLUSIVE PRICE
    28.90€
    27.04€ / paquete de 20
    Ahorro 1.86€
    6% Descuento
En stock
Add to basket
Edge
Add to basket
Excellent for liquid evaporation, weighing solids, heating small amounts of substances and as beaker covers. Rogo Sampaic™ Watch Glasses are manufactured from durable clear borosilicate glass.
  • Available in a range of diameters
  • Corrosion-free and resistant to thermal shock
Specifications
Capacity (Metric) 50 mL
Material Borosilicate Glass
Type Watch Glass
For Use With (Application) Beaker
Temperature Range (Metric) 500 °C
Diameter (Metric) 50 mm