Learn More
Glassco Laboratory Equipments Volumetric Pipettes with Batch Certificate
Product Code. 32210084
Change view
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
Unit Size:
Pack of 15
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
32210084 2 mL Pack of 15
32210055 1 mL Pack of 15
32210062 5 mL Pack of 15
32210068 10 mL Pack of 15
32210054 20 mL Pack of 15
32210022 25 mL Pack of 15
32210052 50 mL Pack of 15
32210058 100 mL Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 32210084 Supplier Glassco Laboratory Equipments Supplier No. EU123.202.02
Fecha estimada de envoi: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
With batch certificate, Class AS.
  • Complies with DIN ISO 648 Standards
  • 1 mark
  • Printed in amber color
  • Chemical resistant
Specifications
Autoclavable Autoclavable
Material Soda Lime Glass
Sterility Non-sterile
Graduated Yes
Tolerance ±0.010 mL
Color Transparent
Volume (Metric) 2 mL
Type Volumetric Pipet
Disposable Reusable
For Use With (Equipment) Pipet Controller
Fit for Purpose Certified Quality
Color Code Orange