missing translation for 'onlineSavingsMsg'
Learn More

Vitronectin Antibody (019), Alexa Fluor™ 594, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-90648AF594

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213276

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Vitronectin Monoclonal specifically detects Vitronectin in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

Vitronectin
Monoclonal
Alexa Fluor 594
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Mouse
Purified
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
19
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
complement S-protein, epibolin, serum spreading factor, Serum-spreading factor, somatomedin B, S-protein, V75, vitronectin, vitronectin (serum spreading factor, somatomedin B, complement S-protein), VN, VNT
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Vitronectin (NP_035837.1; Met1-Lys478).
0.1 mL
Immunology, Innate Immunity
7448
Store at 4°C in the dark.
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales