Learn More
Honeywell™ Vertigo Black PU C&G 1 Gloves
Honeywell™ Vertigo Black PU C&G 1 Gloves for handling of fine products in a dry, slightly greasy or soiled environment.
Marca:  Honeywell™ 2132251-10
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 12605323
Cantidad Precio
1 1.91€ / 1 par
10 1.58€ / 1 par
Stock Remoto 9
Añadir a la cesta
Descripción
Descripción
  • 100% black polyamide knitted glove
  • Black polyurethane coating on palm and fingertips
  • The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
  • The glove offers maximum comfort and added sturdiness
  • The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • The ventilated back and PU coating optimise the glove's breathability
  • The polyamide and its polyurethane coating provide a good abrasion and tear resistance
  • Non pilling, the polyamide fiber is suitable for precision work
  • The gloves are certified as silicone-free to ensure their use in painting environment
  • Its dark color makes it more soilproof, thus ensuring a longer life
Especificaciones
Especificaciones
Polyamide, Polyurethane
10
25 cm
13 gauge
Black
EN 420-2003 Basic standard, EN 388-2016 Mechanical risks