Learn More
3M™ Versaflo™ S-Series Hood Assembly with Inner Collar and Helmet, S-665
Product Code. 31297047 Shop All 3M Products
Change view
Click to view available options
Product Type:
PAPR Assembly with Hood
Product Code. Product Type
31297047 PAPR Assembly with Hood
1 options
This item is not returnable. View return policy
Product Code. 31297047 Supplier 3M™ Supplier No. 7100326013
  • ONLINE EXCLUSIVE PRICE
    290.00€
    193.72€ / paquete de 1
    Ahorro 96.28€
    33% Descuento
Fecha estimada de envoi: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Offer a variety of options for faceseals, collars, shrouds, and suspensions, allowing you to choose the respiratory protection that provides the most value and comfort for your team.
  • Lightweight, versatile and easy to maintain
  • Help protect your workers from a variety of contaminants while maintaining comfort
  • Available in fully assembled integrated hoods for convenience
  • Reusable suspension options offer cost savings
  • Designed to work for your needs
TRUSTED_SUSTAINABILITY
Specifications
Color White
Disposable Single-use
Filter Type Combination
Material Plastic
Product Type PAPR Assembly with Hood
Size One Size
With Exhalation Valve No