Learn More
Binder™ Vacuum Drying Chambers, Series VD
Ensures optimal heat transfer with patented Binder™ expansion shelf technology. Binder™ Vacuum Drying Chamber, Series VD uses homogenous temperature distribution and includes shelves that are easy to position.
8245.00€ - 11405.00€
Productos 2
Código de producto Marca Dimensiones exteriores (longitud x anchura x altura) Precio Cantidad y disponibilidad  
Código de producto Marca Dimensiones exteriores (longitud x anchura x altura) Precio Cantidad y disponibilidad  
15642186 Binder™
9030-0030
8245.00€
cada uno
Fecha estimada de envío
12-02-2019
Añadir a la cesta
15652186 Binder™
9030-0031
11405.00€
cada uno
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • Fast and gentle drying
  • Direct heat transfer through large thermal conducting plates
  • Safe work with one-of-a-kind safety concept
  • Coordinated, modular system with vacuum pump and vacuum module
  • Temperature range: room temperature plus 15 °C to 200 °C
  • APT.line™ preheating chamber technology
  • Controller with time-segment and real-time programming
  • 2 aluminum expansion racks
  • Inert gas connection
  • Shatterproof, spring-mounted safety glass panel
  • Class 2 independent adjustable temperature safety device (DIN 12880) with visual alarm
  • Computer interface: RS 422
  • 2 relay outputs, 24 V DC (max. 0.4 A)