missing translation for 'onlineSavingsMsg'
Learn More

VCIP135/VCPIP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP1-49939

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213281

  • 534.54€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

VCIP135/VCPIP1 Polyclonal specifically detects VCIP135/VCPIP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

VCIP135/VCPIP1
Polyclonal
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
VCPIP1
The immunogen for this product maps to a region between residue 1100 and 1150 of human Valosin-Containing Protein (p97)/p47 Complex-Interacting Protein p135 using the numbering given in entry NP_079330.2 (GeneID 80124).
0.1 mL
Primary
Human
IgG
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunohistochemistry, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:100 - 1:500
DKFZp686G038, DUBA3, EC 3.4.22.-, FLJ23132, KIAA1850FLJ60694, valosin containing protein (p97)/p47 complex interacting protein 1, valosin-containing protein (p97)/p47 complex-interacting protein p135, Valosin-containing protein p97/p47 complex-interacting protein 1, Valosin-containing protein p97/p47 complex-interacting protein p135, VCIP135deubiquitinating protein VCIP135, VCPIP1 VCIP135
Rabbit
Affinity Purified
RUO
80124
Store at 4C. Do not freeze.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only