missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15773-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a homeobox protein and is almost exclusively expressed in the ventral portion of the retina during development. In mouse studies, this gene was found to be required for the correct formation of the optic fissure and other aspects of retinal development.Especificaciones
VAX2 | |
Polyclonal | |
Unconjugated | |
VAX2 | |
DRES93, VAX2, ventral anterior homeobox 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
24113, 25806, 64572 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9JLZ9, Q9UIW0, Q9WTP9 | |
Vax2 | |
VAX2 Fusion Protein Ag8439 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |