Learn More
Thermo Scientific™ Accessories for Varioskan™ LUX Multimode Microplate Reader
Various accessories for use with the Varioskan™ LUX Multimode Microplate Reader.
Marca:  Thermo Scientific™ VLUPD005
Detalles adicionales : Peso : 1.00000kg
 Ver más versiones de este producto
Código de producto. 15310787
  • 5448.00€ / cada uno
Fecha de envío estimada
del proveedor 22-02-2019
Añadir a la cesta
Descripción
Descripción
Varioskan LUX is paired with the powerful fourth generation SkanIt Software, providing excellent usability and flexibility.
  • License free—Each user can install the software on his/her computer
  • Intuitive and flexible interface for multiple users and assays
  • Innovative Virtual Pipetting ToolTM makes it easy to define samples to plate layout
  • Measurement data is continuously saved to the database, so there’s no data lost due to unexpected interruptions such as power outages or accidental aborting
  • Simplified data processing and export
  • Export data in a single mouse click, or have it emailed automatically at the end of the run
  • Several file formats for exporting results: *.xlsx, *.pdf, *.xml, and *.txt
The Varioskan LUX is modular and upgradeable, so you can build the system that best suits your needs. For more information, contact a sales representative.
Especificación
Especificación
Dispenser upgrade for Varioskan LUX
Accessory
Videos
Warranty