Learn More
Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Panel Mounting
VapLock™ Solvent Waste Manifold Caps for Panel Mounting connect to waste containers for a secure closed system
Marca:  Cole-Parmer™ PMP-4
Detalles adicionales : Peso : 0.45000kg
 Ver más versiones de este producto
Código de producto. 15824474
  • 280.91€ / cada uno
Fecha estimada de envío
12-06-2019
Añadir a la cesta
Descripción
Descripción
  • Fits panel aperture of 1-1/4 in and require a 1-3/4 in diameter clearance around the aperture
  • Different materials, numbers and sizes of ports help customize to your application
Warranty
None
Especificaciones
Especificaciones
Solvent Safety
Polypropylene
Solvent Waste Manifold
Solvent-Compatible Containers
Panel Mount
Videos