Learn More
Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys
VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys connect to waste containers for a secure closed system
Marca:  Cole-Parmer™ WM50X-4
Detalles adicionales : Peso : 0.45000kg
 Ver más versiones de este producto
Código de producto. 15824604
  • 461.00€ / cada uno
Fecha estimada de envío
13-12-2019
Añadir a la cesta
Descripción
Descripción
  • Select from a variety of cap sizes to fit your container
  • Different materials, numbers and sizes of ports help customize to your application
Warranty
None
Especificaciones
Especificaciones
Solvent Waste Manifold, PTFE, DIN 50, 1 x 1/2 in. NPTF, 3 x 1/4 in. NPTF; 1/ea
Solvent-Compatible Containers
DIN 50
Solvent Safety
PTFE