missing translation for 'onlineSavingsMsg'
Learn More

VAPB Mouse anti-Human, Clone: 4F6A6, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66191-1-IG-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16817845

  • 319.00€ / 150 microlitros

Envío estimado: 28-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Vesicle-associated membrane protein-associated protein B/C is a protein that in humans is encoded by the VAPB gene. The VAPB gene is found on the 20th human chromosome. The protein encoded by this gene is a type IV membrane protein found in plasma and intracellular vesicle membranes. The encoded protein is found as a homodimer and as a heterodimer with VAPA. This protein also can interact with VAMP1 and VAMP2 and may be involved in vesicle trafficking.
Especificaciones

Especificaciones

VAPB
4F6A6
Unconjugated
VAPB
Liquid
VAPB
VAPB Fusion Protein Ag6313
Protein G
RUO
Antibody
Monoclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95292
ALS8, VAMP associated protein B/C, VAMP B, VAMP B/VAMP C, VAMP C, VAP B, VAP B/VAP C, VAP C, VAPB
Mouse
IgG1
150 μL
-20°C
Primary
9217
Videos
SDS
Documentos
Certificados