missing translation for 'onlineSavingsMsg'
Learn More

VAP-B Antibody (208), Biotin, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-89626B

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18216159

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

VAP-B Monoclonal specifically detects VAP-B in Human samples. It is validated for Western Blot, ELISA.
Especificaciones

Especificaciones

VAP-B
Monoclonal
Biotin
ALS8VAMP-B/VAMP-C, VAMP (vesicle-associated membrane protein)-associated protein B and C, VAMP-associated 33 kDa protein, VAMP-associated protein B/C, VAMP-B, VAP-B, VAP-B/VAP-C, VAPC, VAP-C, vesicle-associated membrane protein-associated protein B/C
This antibody was obtained from a rabbit immunized with purified, recombinant Human VAP-B (O95292-1; Met 1-Pro 132).
0.1 mL
Neuroscience
9217
Store at 4°C in the dark.
IgG
Western Blot, ELISA
208
PBS
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales