missing translation for 'onlineSavingsMsg'
Learn More

Vang-like Protein 1/VANGL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP2-88564

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215838

  • 453.55€ / 100 microlitros
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

Vang-like Protein 1/VANGL1 Polyclonal specifically detects Vang-like Protein 1/VANGL1 in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

Vang-like Protein 1/VANGL1
Polyclonal
Unconjugated
PBS, 2% Sucrose
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
KAI1 C-terminal interacting tetraspanin, Loop-tail protein 2 homolog, LPP2, MGC5338, STB2KITENIN, STBM2, Strabismus 2, Van Gogh-like protein 1, vang (van gogh, Drosophila)-like 1, vang, van gogh-like 1 (Drosophila), vang-like 1 (van gogh, Drosophila), vang-like protein 1
The immunogen is a synthetic peptide directed towards the N-terminal region of Vang-like Protein 1/VANGL1
100 μL
Wnt Signaling Pathway
81839
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales