Learn More
Brady™ Valve Lockout Kit
Product Code. 15623547 Shop All Brady Products
Change view
Click to view available options
Color:
Red/Silver
Unit Size:
Each
Product Code. Color Unit Size
15623547 Red/Silver Each
1 options
This item is not returnable. View return policy
Product Code. 15623547 Supplier Brady™ Supplier No. 806180
Fecha estimada de envoi: 03-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The combination lockout kit gives you everything you need to lock out the most common mechanical and electrical control points.
  • Kit for valve and electrical lockout
Specifications
Alternate Part Number Valve Lockout Kit (NL)
Color Red/Silver
Includes Universal Lockout System - Large, Locking Arm - Large, Cable Fixing + Cable, Adjustable, Gate Valve Casing Universal Cable Locking System + Vinyl Coated Metal Cable (2.44 m), 1 Pk. Brady Security Locks, 1 Pk. Multi Padlock Shackles - 25 mm, Do Not Operate Tag 75 x 160 mm Lockout- shoulder bag, Pen