Learn More
Vadadustat, TRC
Product Code. 30135013
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135013 25 mg 25mg
17910415 50 mg 50mg
17921245 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135013 Supplier Toronto Research Chemicals Supplier No. TRCV84953025MG
 Restricted Item
Fecha estimada de envoi: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1000025-07-9
Molecular Formula C14 H11 Cl N2 O4
Molecular Weight (g/mol) 306.7
Synonym Glycine, N-[[5-(3-chlorophenyl)-3-hydroxy-2-pyridinyl]carbonyl]-, N-[[5-(3-Chlorophenyl)-3-hydroxy-2-pyridinyl]carbonyl]glycine, 2-[[[5-(3-Chlorophenyl)-3-hydroxypyridin-2-yl]carbonyl]amino]acetic acid, AKB 6548, PG 1016548, Vadadustat
IUPAC Name 2-[[5-(3-chlorophenyl)-3-hydroxypyridine-2-carbonyl]amino]acetic acid
SMILES OC(=O)CNC(=O)c1ncc(cc1O)c2cccc(Cl)c2
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C14H11ClN2O4/c15-10-3-1-2-8(4-10)9-5-11(18)13(16-6-9)14(21)17-7-12(19)20/h1-6,18H,7H2,(H,17,21)(H,19,20)
Formula Weight 306.0407
Chemical Name or Material Vadadustat