Descripción
- Simple, compact and sturdy modular design
- Sterilizable, autoclave-compatible
- Table-top or stand-supported device, low space requirement
- Easy disassembly
- Large delivery capacity of 4.4 to 11.6 L/min. with open outlet (the mounting of a valve can reduce the flow rate)
- For air-free, sterile, and inline suspension, emulsifying and deagglomeration
- For vacuum or pressurized operation (up to 6 bar)
- If the DK 25.11 is used, air induction is also prevented in batch operation
- Not self-priming
- A pump can be integrated between intake nozzle and vessel. As a result, viscous fluids can be processed
- Not suitable for continuous operation or cyclical continuous operation
- The hoses used should have an inner diameter size of 13 mm (inlet) and 9 mm (outlet) respectively to fit the DK 25.11 flow chamber
Especificaciones
3000 to 25,000 rpm | |
50/60 Hz | |
13 mm | |
175 mm | |
73 dB |
230 V | |
26 mL | |
450 x 120 x 100 mm | |
Circulation or Flow-Through Processing | |
Swiss |