missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP21 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17856-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the C19 peptidase family, also known as family 2 of ubiquitin carboxy-terminal hydrolases. The encoded protein cleaves ubiquitin from ubiquitinated proteins for recycling in intracellular protein degradation. The encoded protein is also able to release NEDD8, a ubiquitin-like protein, from NEDD8-conjugated proteins. This gene has been referred to as USP16 and USP23 but is now known as USP21. Alternatively spliced transcript variants have been described.Especificaciones
USP21 | |
Polyclonal | |
Unconjugated | |
USP21 | |
Deubiquitinating enzyme 21, Ubiquitin thiolesterase 21, USP16, USP21, USP23 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27005 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UK80 | |
USP21 | |
USP21 Fusion Protein Ag12316 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |