missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19374-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Ubiquitin is a highly conserved protein that is covalently linked to other proteins to regulate their function and degradation. This gene encodes a member of the ubiquitin-specific protease family of cysteine proteases. The enzyme specifically cleaves ubiquitin from ubiquitin-conjugated protein substrates. The protein is found in the nucleus and cytoplasm. It functions as a co-factor of the DNA-bound androgen receptor complex, and is inhibited by a protein in the Ras-GTPase pathway. The human genome contains several pseudogenes similar to this gene.Especificaciones
USP10 | |
Polyclonal | |
Unconjugated | |
USP10 | |
Deubiquitinating enzyme 10, KIAA0190, Ubiquitin thiolesterase 10, UBPO, USP10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22224, 307905, 9100 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P52479, Q14694, Q3KR59 | |
Usp10 | |
USP10 Fusion Protein Ag7233 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |