Learn More
URB754, TRC
Product Code. 30452121
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. Quantity Unit Size
30452121 5 mg 5mg
1 options
This item is not returnable. View return policy
Product Code. 30452121 Supplier Toronto Research Chemicals Supplier No. TRCU8220005MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material URB754
CAS 86672-58-4
Molecular Formula C16 H14 N2 O2
Quantity 5 mg
InChI Formula InChI=1S/C16H14N2O2/c1-10-3-6-12(7-4-10)17-16-18-14-8-5-11(2)9-13(14)15(19)20-16/h3-9H,1-2H3,(H,17,18)
SMILES Cc1ccc(NC2=Nc3ccc(C)cc3C(=O)O2)cc1
IUPAC Name 6-methyl-2-(4-methylanilino)-3,1-benzoxazin-4-one
Molecular Weight (g/mol) 266.29
Formula Weight 266.1055