Learn More
Motic™ Universal Stand, Rectangular Base
Accommodate a variety of different microscopes with this universal microscope stand. Motic™ Universal Stand, Rectangular Base is sturdy and stable.
Marca:  Motic™ 1101000901981
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 11818743
  • 253.00€ / cada uno
Fecha de envío estimada
del proveedor 03-03-2020
Add to basket
Description
Description
  • Suitable for a variety of microscopes.
  • Rectangular base.
  • Stable and sturdy construction.
Specifications
Specifications
Universal
Microscope stand
Rectangular base, 32mm diameter pole
Videos
Documents