Learn More
Corning™ Universal Linear Pipette Stand
Product Code. 16389921 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
Orange
Positions:
4
6
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Positions Unit Size
16389921 Orange 4 Each
16399921 Clear 4 Each
16309931 Clear 6 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16389921 Supplier Corning™ Supplier No. 6066
  • ONLINE EXCLUSIVE PRICE
    65.04€
    58.38€ / paquete de 1
    Ahorro 6.66€
    10% Descuento
Fecha estimada de envoi: 09-12-2026
Add to basket
Add to basket
Bulk Quote Request
This item is not returnable. View return policy
Convenient pipette storage.
Corning™ universal linear stand for four pipettors, (four single-channel, maximum of two 8-channel or 12-channel pipettors)
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Color Orange
Holds Single-channel Pipettes (4) or Multi-channel Pipettes (2)
Material POM
Positions 4
Sterility Non-sterile
For Use With (Equipment) Pipettes
Quantity Case of 1
Length (Metric) 30.6 cm
Product Type Pipette Stand
Height (Metric) 23.8 cm
Width (Metric) 11 cm