missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UNC5C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20240-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The antibody is specific to UNC5C.
This gene product belongs to the UNC-5 family of netrin receptors. Netrins are secreted proteins that direct axon extension and cell migration during neural development. They are bifunctional proteins that act as attractants for some cell types and as repellents for others, and these opposite actions are thought to be mediated by two classes of receptors. The UNC-5 family of receptors mediate the repellent response to netrin; they are transmembrane proteins containing 2 immunoglobulin -like domains and 2 type I thrombospondin motifs in the extracellular region.Especificaciones
UNC5C | |
Polyclonal | |
Unconjugated | |
UNC5C | |
Netrin receptor UNC5C, Protein unc 5 homolog 3, Protein unc 5 homolog C, unc 5 homolog C (C. elegans), UNC5C, UNC5H3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22253, 362049, 8633 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O08747, O95185, Q761X5 | |
Unc5c | |
Peptide | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |