missing translation for 'onlineSavingsMsg'
Learn More

UMP/CMP kinase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11360-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16818383

  • 319.00€ / 150 microlitros

Envío estimado: 27-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

UCK1 (Uridine-cytidine kinase 1) phosphorylates uridine and cytidine to uridine monophosphate and cytidine monophosphate. This enzyme does not phosphorylate deoxyribonucleosides or purine ribonucleosides. Of note, UCK1 is able to use either ATP or GTP as a phosphate donor. UCK1 also possesses the ability to phosphorylate a number of cytidine and uridine nucleoside analogs such as 6-azauridine, 5-fluorouridine, 4-thiouridine, 5-bromouridine, N(4)-acetylcytidine, N(4)-benzoylcytidine, 5-fluorocytidine, 2-thiocytidine, 5-methylcytidine, and N(4)-anisoylcytidine.
Especificaciones

Especificaciones

UMP/CMP kinase
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P30085, Q4KM73, Q9DBP5
CMK, CMPK, CMPK1, Cytidine monophosphate kinase, Cytidylate kinase, Deoxycytidylate kinase, UCK, UMK, UMP CMP kinase, UMP CMPK, UMP/CMP kinase, UMP/CMPK, UMPK, Uridine monophosphate kinase
Rabbit
IgG
150 μL
-20°C
Primary
298410, 51727, 66588
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CMPK1
Liquid
CMPK1
UMP/CMP kinase Fusion Protein Ag1920
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documentos
Certificados