missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UGCGL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14170-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a transcription factor that interacts with specific negative regulatory elements (NREs) to mediate transcriptional repression of certain NK-kappa-B-responsive genes. The protein localizes predomitly to the nucleolus with a small fraction found in the nucleoplasm and cytoplasm.Especificaciones
UGCGL1 | |
Polyclonal | |
Unconjugated | |
UGGT1 | |
GT, HUGT1, UGCGL1, UGGT, UGGT1, UGT1, UGTR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171129, 320011, 56886 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6P5E4, Q9JLA3, Q9NYU2 | |
Uggt1 | |
UGCGL1 Fusion Protein Ag5369 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |