Learn More
Abnova™ UGCG Recombinant Protein
Human UGCG full-length ORF recombinant protein with GST-tag at N-terminal
Marca: Abnova™ H00007357-P01.2ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Glycosphingolipids (GSLs) are a group of membrane components that contain lipid and sugar moieties. They are present in essentially all animal cells and are believed to have important roles in various cellular processes. UDP-glucose ceramide glucosyltransferase catalyzes the first glycosylation step in glycosphingolipid biosynthesis. The product, glucosylceramide, is the core structure of more than 300 GSLs. UGCG is widely expressed and transciption is upregulated during keratinocyte differentiation.
- Molecular Weight: 71.3kDa
- Preparation Method: in vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 Fast Flow
- Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH 8.0 in the elution buffer
Sequence: MALLDLALEGMAVFGFVLFLVLWLMHFMAIIYTRLHLNKKATDKQPYSKLPGVSLLKPLKGVDPNLINNLETFFELDYPKYEVLLCVQDHDDPAIDVCKKLLGKYPNVDARLFIGGKKVGINPKINNLMPGYEVAKYDLIWICDSGIRVIPDTLTDMVNQMTEKVGLVHGLPYVADRQGFAATLEQVYFGTSH
PRYYISANVTGFKCVTGMSCLMRKDVLDQAGGLIAFAQYIAEDYFMAKAIADRGWRFAMSTQVAMQNSGSYSISQFQSRMIRWTKLRINMLPATIICEPISECFVASLIIGWAAHHVFRWDIMVFFMCHCLAWFIFDYIQLRGVQGGTLCFSKLDYAVAWFIRESMTIYIFLSALWDPTISWRTGRYRLRCGGTAEEILDV
Best when used within three months from the date of receipt.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Especificaciones
NP_003349.1 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
69.41 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
2 ug | |
MALLDLALEGMAVFGFVLFLVLWLMHFMAIIYTRLHLNKKATDKQPYSKLPGVSLLKPLKGVDPNLINNLETFFELDYPKYEVLLCVQDHDDPAIDVCKKLLGKYPNVDARLFIGGKKVGINPKINNLMPGYEVAKYDLIWICDSGIRVIPDTLTDMVNQMTEKVGLVHGLPYVADRQGFAATLEQVYFGTSHPRYYISANVTGFKCVTGMSCLMRKDVLDQAGGLIAFAQYIAEDYFMAKAIADRGWRFAMSTQVAMQNSGSYSISQFQSRMIRWTKLRINMLPATIICEPISECFVASLIIGWAAHHVFRWDIMVFFMCHCLAWFIFDYIQLRGVQGGTLCFSKLDYAVAWFIRESMTIYIFLSALWDPTISWRTGRYRLRCGGTAEEILDV | |
GCS | |
UGCG | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
7357 | |
UGCG (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
UGCG | |
Human | |
Recombinant | |
Solution |