missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ UBL3 (Human) Recombinant Protein
Human UBL3 full-length ORF ( AAH59385, 1 a.a. - 117 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00005412-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MSSNVPADMINLRLILVSGKTKEFLFSPNDSASDIAKHVYDNWPMDWEEEQVSSPNILRLIYQGRFLHGNVTLGALKLPFGKTTVMHLVARETLPEPNSQGQRNREKTGESNCCVIL
Especificaciones
AAH59385 | |
ubiquitin-like 3 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
UBL3 | |
GST |
5412 | |
Wheat germ expression system | |
25 μg | |
DKFZp434K151, FLJ32018, HCG-1, PNSC1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |