missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBAP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12385-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Originally named because of its sequence similarity to the Saccharomyces cerevisiae NHP2 (non-histone protein 2), this protein appears to be a highly conserved nuclear protein that is a component of the [U4/U6.U5] tri-snRNP. It binds to the 5′ stem-loop of U4 snRNA. Two transcript variants encoding the same protein have been found for this gene.Especificaciones
UBAP1 | |
Polyclonal | |
Unconjugated | |
UBAP1 | |
NAG20, UAP, UBAP, UBAP 1, UBAP1, ubiquitin associated protein 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
362502, 51271, 67123 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5XIS7, Q8BH48, Q9NZ09 | |
UBAP1 | |
UBAP1 Fusion Protein Ag3055 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |