missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ UBADC1 Recombinant Protein
Marca: Abnova™ H00010422-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Especificaciones
AAH04967.1 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
70.4 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MFVQEEKIFAGKVLRLHICASDGAEWLEEATEDTSVEKLKERCLKHCAHGSLEDPKSITHHKLIHAASERVLSDARTILEENIQDQDVLLLIKKRAPSPLPKMADVSAEEKKKQDQKAPDKEAILRATANLPSYNMDRAAVQTNMRDFQTELRKILVSLIEVAQKLLALNPDAVELFKKANAMLDEDEDERVDEAALRQLTEMGFPENRATKALQLNHMSVPQAMEWLIEHAEDPTIDTPLPGQAPPEAEGATAAASEAAAGASATDEEARDELTEIFKKIRRKREFRADARAVISLMEMGFDEKEVIDALRVNNNQQNAACEWLLGDRKPSPEELDKGIDPDSPLFQAILDNPVVQLGLTNPKTLLAFEDMLENPLNSTQWMNDPETGPVMLQISRIFQTLNRT | |
GBDR1/RP11-432J22.3/UBADC1 | |
UBAC1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
10422 | |
UBADC1 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
UBAC1 | |
Human | |
Recombinant | |
Solution |