missing translation for 'onlineSavingsMsg'
Learn More

U2AF1L4 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 1 publication

Marca:  Novus Biologicals NBP1-06996

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18216054

  • 480.55€ / 0.10 miligramo
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

U2AF1L4 Polyclonal specifically detects U2AF1L4 in Human samples. It is validated for Western Blot, Peptide ELISA.
Especificaciones

Especificaciones

U2AF1L4
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
FLJ35525, MGC33901, splicing factor U2AF 26 kDa subunit, U2 auxiliary factor 26, U2 small nuclear RNA auxiliary factor 1-like 3, U2 small nuclear RNA auxiliary factor 1-like 4, U2 small nuclear RNA auxiliary factor 1-like protein 3, U2 small nuclear RNA auxiliary factor 1-like protein 4, U2(RNU2) small nuclear RNA auxiliary factor 1-like 3, U2(RNU2) small nuclear RNA auxiliary factor 1-like protein 3, U2AF1L3, U2AF1L3V1, U2AF1-like protein 3, U2AF1RS3, U2AF1-RS3, U2af26
Goat
Affinity Purified
RUO
199746
Human
IgG
Western Blot, ELISA
0.5 mg/mL
Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:32000
NP_001035515.1
U2AF1L4
Peptide with sequence C-EDGERAVAELSNR corresponding to internal region according to NP_659424.2.
0.1 mg
Primary
This antibody is expected to recognize both reported isoforms (NP_001035515.1 and NP_659424.2). No cross-reactivity with U2AF1 expected.
Store at -20C. Avoid freeze-thaw cycles.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only