missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TYMS Mouse anti-Human, Clone: 1F5B7, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66725-1-IG-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Thymidylate Synthase (TS) (EC 2.1.1.45) converts deoxyuridine monophosphate (dUMP) to deoxythymidine monophosphate (dTMP), which is essential for DNA biosynthesis. TS is a critical target for the fluoropyrimidines, an important group of antineoplastic drugs that are widely used in the treatment of solid tumors. Both 5-FU and fluorodeoxyuridine are converted in tumor cells to FdUMP which inactivates TS by formation of a ternary covalent complex in the presence of the folate cofactor 5,10-methylenetetrahydrofolate. Expression of TS protein is associated with response to 5-fluorouracil (5-FU) in human colorectal, gastric, head and neck, and breast carcinomas.Especificaciones
TYMS | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P04818 | |
TYMS | |
TYMS Fusion Protein Ag7027 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1F5B7 | |
Unconjugated | |
TYMS | |
HsT422, Thymidylate synthase, thymidylate synthetase, TMS, TS, TSase, TYMS | |
Mouse | |
Protein G | |
RUO | |
7298 | |
-20°C | |
Liquid |