missing translation for 'onlineSavingsMsg'
Learn More

TYKi Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody

Marke:  Novus Biologicals NBP2-82045IR

Weitere Details : Peso : 0.00970kg

Artikelnummer. 18216327

  • 577.74€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Beschreibung

Beschreibung

TYKi Polyclonal antibody specifically detects TYKi in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Spezifikation

Spezifikation

TYKi
Polyclonal
50mM Sodium Borate
Rabbit
Peptide affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, ELISA, Immunohistochemistry
DyLight 755
cytidine monophosphate (UMP-CMP) kinase 2, mitochondrial, cytidylate kinase 2, EC 2.7.4.14, mitochondrial, mitochondrial UMP-CMP kinase, thymidine monophosphate kinase 2, thymidylate kinase family LPS-inducible member, TMPK2, TYKi, UMP-CMPK2
TYKi antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human TYKi. The immunogen is located within amino acids 370 - 420 of TYKi. Amino Acid Squence: PEERLQRLQGRGMEKTREE
0.1 mL
Protein Kinase
129607
Store at 4°C in the dark.
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote