Learn More
WTW™ Turb™ Turbidity Standard
Product Code. 11725156
Change view
Click to view available options
Turbidity:
10,000 NTU
Unit Size:
Each
Product Code. Turbidity Unit Size
11725156 10,000 NTU Each
1 options
This item is not returnable. View return policy
Product Code. 11725156 Supplier WTW™ Supplier No. 600546
 Restricted Item
Fecha estimada de envoi: 29-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
AMCO Clear™ standards are polymer calibration standards for Turb™ meters and are much more accurate and stable without drifting than Formazine
AMCO Clear™ standards are polymer calibration standards for Turb™ meters and are much more accurate and stable without drifting than Formazine. They are traceable to Formazine according to N.I.S.T and are lot-certified. The long-term stability without particle change in size and shape and the high manufacturing tolerance of only ±1% helps achieve the highest results accuracy. The standards are adapted to the respective device optics in their particle composition. The natural drifting of Formazine and the much higher manufacturing tolerance with Formazine grants them clear advantages. This makes AMCO Clear™ standards especially suitable for applications in the lower measuring range such as drinking water.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Turb 555 WL Laboratory Turbidimeter
Description Turbidity standard
Turbidity 10,000 NTU