Learn More
Polvo de tungsteno, 99,95% (base metálica), Thermo Scientific Chemicals
Catalog Number 11318998
Click to view available options
Cantidad:
100 g
500 g
2 kg
7440-33-7
W
183.84
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
23964
CHEBI:27998
tungsteno
[W]
7440-33-7
W
183.84
MFCD00011461
WFKWXMTUELFFGS-UHFFFAOYSA-N
wolfram, ion, va, tungsten, elemental, wolframium, unii-v9306cxo6g, 4+ ion, tungsten, soluble compounds, tungsten, insoluble compounds, powder
23964
CHEBI:27998
tungsteno
[W]
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Tungsten
Color Gris
Aspecto Light Gray powder
Intervalo de porcentaje de ensayo 99.95% (Metals basis)
Cantidad 2 kg
Tamaño de malla -200+325 Mesh
Formulario Polvo