Learn More
Gilac™ Tumblers, 22 Cycle, White
Product Code. 17938631
Change view
Click to view available options
Capacity (Metric):
0.22 L
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
17938631 0.22 L Each
1 options
This item is not returnable. View return policy
Product Code. 17938631 Supplier Gilac™ Supplier No. G627221
  • ONLINE EXCLUSIVE PRICE
    10.85€
    7.17€ / paquete de 1
    Ahorro 3.68€
    33% Descuento
Fecha estimada de envoi: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Reusable plastic 22 Cycle Tumblers with outstanding durability even in the intensive use cycle.
  • Reusable plastic tumbler.
  • Outstanding durability even in the intensive use cycle.
Specifications
Material Polypropylene
Capacity (Metric) 0.22 L
Capacity (English) 7.4 oz.
Quantity 10 Pk.
Product Type Tumbler Cup