Learn More
Tulobuterol Hydrochloride, 98%, Thermo Scientific Chemicals
Product Code. 15434049
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
15434049 1 g 1g
1 options
This item is not returnable. View return policy
Product Code. 15434049 Supplier Thermo Scientific Alfa Aesar Supplier No. J61448.03
 Restricted Item
En stock
Add to basket
This item is not returnable. View return policy
beta-2-adrenergic receptor agonist | CAS: 56776-01-3 | C12H18ClNO·HCl
beta-2-adrenergic receptor agonist
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 56776-01-3
Molecular Formula C12H18ClNO·HCl
Specifications
Quantity 1 g
Description White
Content And Storage 2°C to 8°C
Format Powder
For Use With (Application) beta-2-adrenergic receptor agonist
Formula Weight 264.19
Sensitivity Hygroscopic
Melting Point 159°C to 163°C
Solubility Soluble in methanol or water
Product Type Signal Transduction Reagent
Form Powder
Assay 98%