Learn More
Cytiva Tube Holder AKTA™ avant polyoxymethylene
Product Code. 12475610 Shop All Cytiva Products
Change view
Click to view available options
Material:
Polyoxymethylene (POM-C)
Unit Size:
Pack of 5
Product Code. Material Unit Size
12475610 Polyoxymethylene (POM-C) Pack of 5
1 options
This item is not returnable. View return policy
Product Code. 12475610 Supplier Cytiva Supplier No. 28954329
  • ONLINE EXCLUSIVE PRICE
    105.08€
    67.38€ / paquete de 5
    Ahorro 37.70€
    35% Descuento
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
For the organisation of small tubing on the wet side of ÅKTA avant or ÅKTA pure systems. Attached to the system by small holes along left edge of wet side and on pump fronts.
TRUSTED_SUSTAINABILITY
Specifications
Description Tube Holder
For Use With (Application) Organize small tubing on the wet side of AKTA avant or AKTA pure
Material Polyoxymethylene (POM-C)
Quantity Pk. of 5